accessibility menu, dialog, popup

UserName

CAS RN 6342-77-4

IUPAC Name: 3-(2-methoxyphenyl)propanoic acid
Synonyms: 3-(2-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: XSZSNLOPIWWFHS-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1CCC(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results