missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenyl)-1H-pyrazole, 97%

Catalog No. AAH3449203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3449203 1 g
AAH3449206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3449203 Supplier Thermo Scientific Chemicals Supplier No. H3449203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59843-63-9 | C10H10N2O | 174.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59843-63-9
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD02091524
InChI Key KLPGJCMICASHKV-UHFFFAOYSA-N
Synonym 3-2-methoxyphenyl-1h-pyrazole, 5-2-methoxyphenyl-1h-pyrazole, 1h-pyrazole, 3-2-methoxyphenyl, 3-2-methoxy-phenyl-1h-pyrazole, 3-2-methoxyphenyl pyrazole, 2-1h-pyrazol-3-yl anisole, 2-methoxy-1-pyrazol-3-ylbenzene
PubChem CID 2736764
IUPAC Name 5-(2-methoxyphenyl)-1H-pyrazole
SMILES COC1=CC=CC=C1C2=CC=NN2

Specifications

Melting Point 82°C to 84°C
Quantity 1 g
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material 3-(2-Methoxyphenyl)-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only