CAS RN 59843-63-9
IUPAC Name:
5-(2-methoxyphenyl)-1H-pyrazole
Synonyms:
3-(2-methoxyphenyl)-2H-pyrazole
Molecular Weight (g/mol):
174.2
Molecular Formula:
C10H10N2O
InChi Key:
KLPGJCMICASHKV-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1C1=CC=NN1
1
–
1
of
1
results
1
Promotions Available
3-(2-Methoxyphenyl)-1H-pyrazole, 97%
3-(2-Methoxyphenyl)-1H-pyrazole, 97%
CAS: 59843-63-9 Molecular Formula: C10H10N2O Molecular Weight (g/mol): 174.203 MDL Number: MFCD02091524 InChI Key: KLPGJCMICASHKV-UHFFFAOYSA-N Synonym: 3-2-methoxyphenyl-1h-pyrazole,5-2-methoxyphenyl-1h-pyrazole,1h-pyrazole, 3-2-methoxyphenyl,3-2-methoxy-phenyl-1h-pyrazole,3-2-methoxyphenyl pyrazole,2-1h-pyrazol-3-yl anisole,2-methoxy-1-pyrazol-3-ylbenzene PubChem CID: 2736764 IUPAC Name: 5-(2-methoxyphenyl)-1H-pyrazole SMILES: COC1=CC=CC=C1C2=CC=NN2