missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Fluorophenyl)-1H-pyrazole, 98%

Catalog No. p-7032104
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3345003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3345003 Supplier Thermo Scientific Chemicals Supplier No. H3345003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149739-32-2 | C9H7FN2 | 162.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149739-32-2
Molecular Formula C9H7FN2
Molecular Weight (g/mol) 162.17
MDL Number MFCD00973917
InChI Key PZNGOFHHOGORBU-UHFFFAOYSA-N
Synonym 3-2-fluorophenyl-1h-pyrazole, 3-2-fluorophenyl pyrazole, 1h-pyrazole, 3-2-fluorophenyl, 1h-pyrazole,3-2-fluorophenyl, 5-2-fluorophenyl-1h-pyrazole, 3-2-fluoro-phenyl-1h-pyrazole, 3-2-fluorophenyl-2h-pyrazole
PubChem CID 2774734
IUPAC Name 5-(2-fluorophenyl)-1H-pyrazole
SMILES FC1=CC=CC=C1C1=CC=NN1

Specifications

Melting Point 58°C to 61°C
Quantity 1 g
Formula Weight 162.17
Percent Purity 98%
Chemical Name or Material 3-(2-Fluorophenyl)-1H-pyrazole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only