accessibility menu, dialog, popup

UserName

CAS RN 149739-32-2

IUPAC Name: 5-(2-fluorophenyl)-1H-pyrazole
Synonyms: 3-(2-fluorophenyl)-2H-pyrazole
Molecular Weight (g/mol): 162.17
Molecular Formula: C9H7FN2
InChi Key: PZNGOFHHOGORBU-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results