missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%

CAS: 1760-24-3 | C8H22N2O3Si | 222.36 g/mol

Supplier:  Thermo Scientific Chemicals A169810B

 View more versions of this product

Catalog No. AAA169810B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

261°C to 263°C
1760-24-3
C8H22N2O3Si
UN2735
136°C (276°F)
PHQOGHDTIVQXHL-UHFFFAOYSA-N
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
15659
90%
1.442
1.01
3-(2-Aminoethylamino)propyltrimethoxysilane
1000 g
MFCD00008173
636230
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
CO[Si](CCCNCCN)(OC)OC
222.36
222.36
Technical
Air and moisture sensitive

Chemical Identifiers

1760-24-3
222.36
PHQOGHDTIVQXHL-UHFFFAOYSA-N
15659
CO[Si](CCCNCCN)(OC)OC
C8H22N2O3Si
MFCD00008173
n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c

H315-H317-H318

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 217-164-6

RTECSNumber : KV7400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only