missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%

Catalog No. p-7023429
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1698118 50 g
AAA1698130 250 g
AAA169810B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1698118 Supplier Thermo Scientific Chemicals Supplier No. A1698118
Only null left

CAS: 1760-24-3 | C8H22N2O3Si | 222.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1760-24-3
Molecular Formula C8H22N2O3Si
Molecular Weight (g/mol) 222.36
MDL Number MFCD00008173
InChI Key PHQOGHDTIVQXHL-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl ethylenediamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 3-2-aminoethylamino propyltrimethoxysilane, en-aptas, silicone a-1120, prosil 3128, aas-m, n1-3-trimethoxysilyl propyl ethane-1,2-diamine, unii-28zcs5ga8g, 3-2-aminoethyl aminopropyl trimethoxysilane
PubChem CID 15659
IUPAC Name N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine
SMILES CO[Si](CCCNCCN)(OC)OC

Specifications

Boiling Point 261°C to 263°C
Quantity 50 g
UN Number UN2735
Beilstein 636230
Flash Point 136°C (276°F)
Formula Weight 222.36
Percent Purity 90%
Grade Technical
Refractive Index 1.442
Sensitivity Air and moisture sensitive
Density 1.01
Chemical Name or Material 3-(2-Aminoethylamino)propyltrimethoxysilane
Show More Show Less
Hazard Category H315-H317-H318
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 217-164-6
RTECSNumber KV7400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only