CAS RN 1760-24-3
3-(2-Aminoethylamino)propyltrimethoxysilane, 96%
CAS: 1760-24-3 Molecular Formula: C8H22N2O3Si Molecular Weight (g/mol): 222.36 MDL Number: MFCD00008173 InChI Key: PHQOGHDTIVQXHL-UHFFFAOYSA-N Synonym: n-3-trimethoxysilyl propyl ethylenediamine,n-2-aminoethyl-3-aminopropyltrimethoxysilane,3-2-aminoethylamino propyltrimethoxysilane,en-aptas,silicone a-1120,prosil 3128,aas-m,n1-3-trimethoxysilyl propyl ethane-1,2-diamine,unii-28zcs5ga8g,3-2-aminoethyl aminopropyl trimethoxysilane PubChem CID: 15659 IUPAC Name: N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine SMILES: CO[Si](CCCNCCN)(OC)OC
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%
CAS: 1760-24-3 Molecular Formula: C8H22N2O3Si Molecular Weight (g/mol): 222.36 MDL Number: MFCD00008173 InChI Key: PHQOGHDTIVQXHL-UHFFFAOYSA-N Synonym: n-3-trimethoxysilyl propyl ethylenediamine,n-2-aminoethyl-3-aminopropyltrimethoxysilane,3-2-aminoethylamino propyltrimethoxysilane,en-aptas,silicone a-1120,prosil 3128,aas-m,n1-3-trimethoxysilyl propyl ethane-1,2-diamine,unii-28zcs5ga8g,3-2-aminoethyl aminopropyl trimethoxysilane PubChem CID: 15659 IUPAC Name: N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine SMILES: CO[Si](CCCNCCN)(OC)OC
3-(2-Aminoethylamino)propyltrimethoxysilane, tech. 90%
CAS: 1760-24-3 Molecular Formula: C8H22N2O3Si Molecular Weight (g/mol): 222.36 MDL Number: MFCD00008173 InChI Key: PHQOGHDTIVQXHL-UHFFFAOYSA-N Synonym: n-3-trimethoxysilyl propyl ethylenediamine,n-2-aminoethyl-3-aminopropyltrimethoxysilane,3-2-aminoethylamino propyltrimethoxysilane,en-aptas,silicone a-1120,prosil 3128,aas-m,n1-3-trimethoxysilyl propyl ethane-1,2-diamine,unii-28zcs5ga8g,3-2-aminoethyl aminopropyl trimethoxysilane PubChem CID: 15659 IUPAC Name: N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine SMILES: CO[Si](CCCNCCN)(OC)OC
3-(2-Aminoethylamino)propyltrimethoxysilane 97.0+%, TCI America™
CAS: 1760-24-3 Molecular Formula: C8H22N2O3Si Molecular Weight (g/mol): 222.36 MDL Number: MFCD00008173 InChI Key: PHQOGHDTIVQXHL-UHFFFAOYSA-N Synonym: n-3-trimethoxysilyl propyl ethylenediamine,n-2-aminoethyl-3-aminopropyltrimethoxysilane,3-2-aminoethylamino propyltrimethoxysilane,en-aptas,silicone a-1120,prosil 3128,aas-m,n1-3-trimethoxysilyl propyl ethane-1,2-diamine,unii-28zcs5ga8g,3-2-aminoethyl aminopropyl trimethoxysilane PubChem CID: 15659 IUPAC Name: N'-(3-trimethoxysilylpropyl)ethane-1,2-diamine SMILES: CO[Si](CCCNCCN)(OC)OC