missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-1-yl)aniline, 97%

CAS: 14213-12-8 | C7H7N5 | 161.168 g/mol

Supplier:  Thermo Scientific Chemicals H6382906

 View more versions of this product

Catalog No. AAH6382906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(1H-Tetrazol-1-yl)aniline
C7H7N5
5 g
YXWXLBHIUVBLIH-UHFFFAOYSA-N
3-(tetrazol-1-yl)aniline
863007
97%
14213-12-8
MFCD06687920
3-1h-tetrazol-1-yl aniline, 3-tetrazol-1-yl-phenylamine, 3-1h-tetrazol-1-yl benzenamine, 3-1,2,3,4-tetrazol-1-yl aniline, 3-1h-1,2,3,4-tetrazol-1-yl aniline, 3-tetrazol-1-yl phenylamine, 3-1,2,3,4-tetraazolyl phenylamine, acmc-209cnn, 3-tetrazol-1-yl aniline, 3-tetrazol-1-ylphenylamine
C1=CC(=CC(=C1)N)N2C=NN=N2
161.168
161.16
Solid

Chemical Identifiers

14213-12-8
161.168
YXWXLBHIUVBLIH-UHFFFAOYSA-N
863007
C1=CC(=CC(=C1)N)N2C=NN=N2
C7H7N5
MFCD06687920
3-1h-tetrazol-1-yl aniline, 3-tetrazol-1-yl-phenylamine, 3-1h-tetrazol-1-yl benzenamine, 3-1,2,3,4-tetrazol-1-yl aniline, 3-1h-1,2,3,4-tetrazol-1-yl aniline, 3-tetrazol-1-yl phenylamine, 3-1,2,3,4-tetraazolyl phenylamine, acmc-209cnn, 3-tetrazol-1-yl aniline, 3-tetrazol-1-ylphenylamine
3-(tetrazol-1-yl)aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only