missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-1-yl)aniline, 97%

Catalog No. p-7055292
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63829MD 250 mg
AAH6382903 1 g
AAH6382906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63829MD Supplier Thermo Scientific Chemicals Supplier No. H63829MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14213-12-8 | C7H7N5 | 161.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14213-12-8
Molecular Formula C7H7N5
Molecular Weight (g/mol) 161.168
MDL Number MFCD06687920
InChI Key YXWXLBHIUVBLIH-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-1-yl aniline, 3-tetrazol-1-yl-phenylamine, 3-1h-tetrazol-1-yl benzenamine, 3-1,2,3,4-tetrazol-1-yl aniline, 3-1h-1,2,3,4-tetrazol-1-yl aniline, 3-tetrazol-1-yl phenylamine, 3-1,2,3,4-tetraazolyl phenylamine, acmc-209cnn, 3-tetrazol-1-yl aniline, 3-tetrazol-1-ylphenylamine
PubChem CID 863007
IUPAC Name 3-(tetrazol-1-yl)aniline
SMILES C1=CC(=CC(=C1)N)N2C=NN=N2

Specifications

Quantity 250 mg
Formula Weight 161.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-(1H-Tetrazol-1-yl)aniline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only