missing translation for 'onlineSavingsMsg'
Learn More

[3-(1H-Pyrazol-1-yl)phenyl]methanol, ≥90%, Thermo Scientific™

Catalog No. CC32909CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC32909CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC32909CB Supplier Thermo Scientific Chemicals Supplier No. CC32909CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 864068-80-4
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.20
MDL Number MFCD06740322
InChI Key VINVOQJANISHSK-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-yl phenyl methanol, 3-1-pyrazolyl benzyl alcohol, 3-pyrazol-1-yl-phenyl-methanol, 3-pyrazol-1-yl phenyl methanol, 3-pyrazol-1-ylphenyl methanol, 3-pyrazolylphenyl methan-1-ol, 3-1h-pyrazole-1-yl benzyl alcohol, benzenemethanol, 3-1h-pyrazol-1-yl, benzenemethanol,3-1h-pyrazol-1-yl
PubChem CID 7164580
IUPAC Name [3-(1H-pyrazol-1-yl)phenyl]methanol
SMILES OCC1=CC(=CC=C1)N1C=CC=N1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 174.2
Percent Purity 90%
Chemical Name or Material [3-(1H-pyrazol-1-yl)phenyl]methanol
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.