CAS RN 864068-80-4
IUPAC Name:
[3-(1H-pyrazol-1-yl)phenyl]methanol
Synonyms:
[3-(pyrazol-1-yl)phenyl]methanol
Molecular Weight (g/mol):
174.2
Molecular Formula:
C10H10N2O
InChi Key:
VINVOQJANISHSK-UHFFFAOYSA-N
SMILES:
OCC1=CC(=CC=C1)N1C=CC=N1
1
–
2
of
2
results
1
[3-(1H-Pyrazol-1-yl)phenyl]methanol, ≥90%, Thermo Scientific™
CAS: 864068-80-4 Molecular Formula: C10H10N2O Molecular Weight (g/mol): 174.20 MDL Number: MFCD06740322 InChI Key: VINVOQJANISHSK-UHFFFAOYSA-N Synonym: 3-1h-pyrazol-1-yl phenyl methanol,3-1-pyrazolyl benzyl alcohol,3-pyrazol-1-yl-phenyl-methanol,3-pyrazol-1-yl phenyl methanol,3-pyrazol-1-ylphenyl methanol,3-pyrazolylphenyl methan-1-ol,3-1h-pyrazole-1-yl benzyl alcohol,benzenemethanol, 3-1h-pyrazol-1-yl,benzenemethanol,3-1h-pyrazol-1-yl PubChem CID: 7164580 IUPAC Name: [3-(1H-pyrazol-1-yl)phenyl]methanol SMILES: OCC1=CC(=CC=C1)N1C=CC=N1