missing translation for 'onlineSavingsMsg'
Learn More

[3-(1H-1,2,4-Triazol-1-ylmethyl)phenyl]methanol, ≥97%, Thermo Scientific™

Catalog No. CC45409CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC45409CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC45409CB Supplier Thermo Scientific Chemicals Supplier No. CC45409CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 871825-54-6
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.22
MDL Number MFCD08271929
InChI Key PZRKSBSNVMSIJI-UHFFFAOYSA-N
Synonym 3-1h-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl phenyl methanol, 3-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl benzyl alcohol, 3-1,2,4-triazolylmethyl phenyl methan-1-ol
PubChem CID 18525775
SMILES OCC1=CC=CC(CN2C=NC=N2)=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
IUPAC Name [3-(1,2,4-triazol-1-ylmethyl)phenyl]methanol
Formula Weight 189.22
Percent Purity 95%
Chemical Name or Material [3-(1H-1, 2, 4-Triazol-1-ylmethyl)phenyl]methanol
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.