missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Naphthyl)-L-alanine, 95%
CAS: 55516-54-6 | C13H13NO2 | 215.25 g/mol
$96.21 - $402.49
Chemical Identifiers
| CAS | 55516-54-6 |
|---|---|
| Molecular Formula | C13H13NO2 |
| Molecular Weight (g/mol) | 215.25 |
| MDL Number | MFCD00066086 |
| InChI Key | OFYAYGJCPXRNBL-LBPRGKRZSA-N |
| Synonym | l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine |
| PubChem CID | 2724883 |
| SMILES | [NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6358603
|
Thermo Scientific Chemicals
H6358603 |
1 g |
Each for $96.21
|
|
|||||
|
AAH6358606
|
Thermo Scientific Chemicals
H6358606 |
5 g |
Each for $402.49
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 55516-54-6 | |
| 215.25 | |
| OFYAYGJCPXRNBL-LBPRGKRZSA-N | |
| 2724883 |
| C13H13NO2 | |
| MFCD00066086 | |
| l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine | |
| [NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O |
Specifications
| 55516-54-6 | |
| MFCD00066086 | |
| l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine | |
| OFYAYGJCPXRNBL-LBPRGKRZSA-N | |
| (2S)-2-azaniumyl-3-(naphthalen-1-yl)propanoate | |
| 215.25 | |
| 215.25 | |
| Solid |
| C13H13NO2 | |
| 4352833 | |
| Slightly soluble in water. | |
| [NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O | |
| 1 g | |
| 2724883 | |
| 95% | |
| 3-(1-Naphthyl)-L-alanine |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only