missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)-L-alanine, 95%

CAS: 55516-54-6 | C13H13NO2 | 215.25 g/mol

Supplier:  Thermo Scientific Chemicals H6358606

 View more versions of this product

Catalog No. AAH6358606


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

3-(1-Naphthyl)-L-alanine
C13H13NO2
4352833
Slightly soluble in water.
[NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O
5 g
2724883
95%
55516-54-6
MFCD00066086
l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine
OFYAYGJCPXRNBL-LBPRGKRZSA-N
(2S)-2-amino-3-naphthalen-1-ylpropanoic acid
215.25
215.25
Solid

Chemical Identifiers

55516-54-6
215.25
OFYAYGJCPXRNBL-LBPRGKRZSA-N
2724883
C13H13NO2
MFCD00066086
l-1-naphthylalanine, 3-1-naphthyl-l-alanine, h-1-nal-oh, s-2-amino-3-naphthalen-1-yl propanoic acid, 3-1-naphthyl-l-ala, unii-9g930ul92r, naphthalen-2-yl-3-alanine, 2s-2-amino-3-naphthalen-1-yl propanoic acid, 3-naphth-1-yl-d-alanine, 1-naphthyl-l-alanine
[NH3+][C@@H](CC1=C2C=CC=CC2=CC=C1)C([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only