missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2E,4E)-5-Phenyl-2,4-pentadienoic Acid 98.0+%, TCI America™
Supplier: TCI America P24225G
Specifications
| (2E,4E)-5-Phenyl-2,4-pentadienoic Acid | |
| 168°C | |
| C11H10O2 | |
| 5 g | |
| FEIQOMCWGDNMHM-KBXRYBNXSA-N | |
| (2E,4E)-5-phenylpenta-2,4-dienoic acid | |
| 1549512 | |
| ≥98.0% (GC,T) |
| 28010-12-0 | |
| White | |
| MFCD00014018 | |
| (E,E)-Cinnamylideneacetic Acid | |
| C1=CC=C(C=C1)C=CC=CC(=O)O | |
| 174.199 | |
| 174.20 | |
| Crystalline Powder |
Chemical Identifiers
| 28010-12-0 | |
| 174.199 | |
| FEIQOMCWGDNMHM-KBXRYBNXSA-N | |
| 1549512 | |
| C1=CC=C(C=C1)C=CC=CC(=O)O |
| C11H10O2 | |
| MFCD00014018 | |
| (E,E)-Cinnamylideneacetic Acid | |
| (2E,4E)-5-phenylpenta-2,4-dienoic acid |
Safety and Handling
TSCA : No