missing translation for 'onlineSavingsMsg'
Learn More

(2E,4E)-5-Phenyl-2,4-pentadienoic Acid 98.0+%, TCI America™

$348.46 - $1389.97

Chemical Identifiers

CAS 28010-12-0
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00014018
InChI Key FEIQOMCWGDNMHM-KBXRYBNXSA-N
Synonym (E,E)-Cinnamylideneacetic Acid
PubChem CID 1549512
IUPAC Name (2E,4E)-5-phenylpenta-2,4-dienoic acid
SMILES C1=CC=C(C=C1)C=CC=CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P24225G
View Documents
TCI America
P24225G
5 g
Each for $348.46
Only null left
 
P242225G
View Documents
TCI America
P242225G
25 g
Each for $1,389.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

28010-12-0
174.199
FEIQOMCWGDNMHM-KBXRYBNXSA-N
1549512
C1=CC=C(C=C1)C=CC=CC(=O)O
C11H10O2
MFCD00014018
(E,E)-Cinnamylideneacetic Acid
(2E,4E)-5-phenylpenta-2,4-dienoic acid

Specifications

28010-12-0
White
MFCD00014018
(E,E)-Cinnamylideneacetic Acid
C1=CC=C(C=C1)C=CC=CC(=O)O
174.199
174.20
Crystalline Powder
168°C
C11H10O2
5 g
FEIQOMCWGDNMHM-KBXRYBNXSA-N
(2E,4E)-5-phenylpenta-2,4-dienoic acid
1549512
≥98.0% (GC,T)
(2E,4E)-5-Phenyl-2,4-pentadienoic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'