missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 5% (w/v) Aqueous Solution, Spectrum™ Chemical

Catalog No. 18600203
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600203 4 L Poly Bottle
18600202 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600203 Supplier Spectrum Chemical Mfg Cor Supplier No. A1014LTPL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A-101, 64-19-7

Spectrum™ Chemical Acetic Acid, 5 Percent (w/v) Aqueous Solution is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak Acid, when concentrated it is corrosive and can attack the skin. A useful chemical reagent for the production of chemical compounds, Acetic acid has many uses. Its main use resides in the production of vinyl acetate monomer, but it is also used in the production of ester and acetic anhydride. As a solvent it is used in the purification by recrystallization of organic compounds and in the production of terephtalic acid.

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 5%, 95%
Quantity 4 L
Packaging Poly Bottle
Percent Purity 5% ±0.05%
Grade Chemical Solution