Learn More
Justrite™ 23 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 23 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Self-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
$4,539.40 - $4,713.96
Description
- Keep personnel and parts away from fluids and contaminants
- Durable, anti-slip grates help keep personnel and other objects above contaminated wash down water
- Made of chemical resistant, easy-to-clean polyethylene
- Service temperature range: -30F to 120F
Specifications
| Self-close | |
| 23 gal. |