Learn More
Restek™ 20 mm Pressure Release Aluminum Seals with Septa, Assembled DFS Item
Preassembled seals save time and increase productivity
Supplier:  Restek™ RK21766
 View more versions of this product
Catalog No. 06718655
Only null left
Explore available promotions
Description
Description
  • Color: Silver
  • Material: PTFE/Gray Butyl Rubber Septa
Specifications
Specifications
Silver
PTFE, Butyl Rubber
Crimp Cap
Aluminum
1000
20 mm
Product Suggestions
Product Suggestions