missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Undecanol 98.0+%, TCI America™
$60.75 - $144.30
Chemical Identifiers
| CAS | 1653-30-1 |
|---|---|
| Molecular Formula | C11H24O |
| Molecular Weight (g/mol) | 172.31 |
| MDL Number | MFCD00021958 |
| InChI Key | XMUJIPOFTAHSOK-UHFFFAOYNA-N |
| Synonym | 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol |
| PubChem CID | 15448 |
| ChEBI | CHEBI:77930 |
| IUPAC Name | undecan-2-ol |
| SMILES | CCCCCCCCCC(C)O |
Chemical Identifiers
| C11H24O | |
| MFCD00021958 | |
| 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol | |
| CHEBI:77930 | |
| CCCCCCCCCC(C)O |
Specifications
| 1653-30-1 | |
| Colorless | |
| C11H24O | |
| 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol | |
| CCCCCCCCCC(C)O | |
| 172.31 | |
| CHEBI:77930 | |
| ≥98.0% (GC) | |
| 2-Undecanol |
| 3°C | |
| 228°C | |
| MFCD00021958 | |
| XMUJIPOFTAHSOK-UHFFFAOYNA-N | |
| undecan-2-ol | |
| 15448 | |
| 172.31 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217&(2)-3704
TSCA : Yes