missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Undecanol 98.0+%, TCI America™
Supplier: TCI America U002725ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| C11H24O | |
| MFCD00021958 | |
| 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol | |
| CHEBI:77930 | |
| CCCCCCCCCC(C)O |
Specifications
| 2-Undecanol | |
| 3°C | |
| 228°C | |
| 25 mL | |
| 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol | |
| CCCCCCCCCC(C)O | |
| 172.31 | |
| CHEBI:77930 | |
| ≥98.0% (GC) |
| 1653-30-1 | |
| Colorless | |
| C11H24O | |
| MFCD00021958 | |
| XMUJIPOFTAHSOK-UHFFFAOYNA-N | |
| undecan-2-ol | |
| 15448 | |
| 172.31 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217&(2)-3704
TSCA : Yes