missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanol 98.0+%, TCI America™

Catalog No. U00275ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00275ML 5 mL
U002725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00275ML Supplier TCI America Supplier No. U00275ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1653-30-1
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00021958
InChI Key XMUJIPOFTAHSOK-UHFFFAOYNA-N
Synonym 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
PubChem CID 15448
ChEBI CHEBI:77930
IUPAC Name undecan-2-ol
SMILES CCCCCCCCCC(C)O

Specifications

Melting Point 3°C
Color Colorless
Boiling Point 228°C
Quantity 5 mL
Formula Weight 172.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Undecanol
EINECSNumber (2)-0217&(2)-3704
TSCA Yes