missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanol, 98+%

CAS: 1653-30-1 | C11H24O | 172.31 g/mol

$130.41 - $809.55

Chemical Identifiers

CAS 1653-30-1
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00021958
InChI Key XMUJIPOFTAHSOK-UHFFFAOYNA-N
Synonym 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
PubChem CID 15448
ChEBI CHEBI:77930
IUPAC Name undecan-2-ol
SMILES CCCCCCCCCC(C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1444109
View Documents Promotion Details
N/A 10 g
Each for $130.41
Only null left
 
AAA1444118
View Documents Promotion Details
N/A 50 g
Each for $400.42
Only null left
 
AAA1444130
View Documents Promotion Details
N/A 250 g
Each for $809.55
Only null left
 
Description

Description

It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

1653-30-1
172.31
XMUJIPOFTAHSOK-UHFFFAOYNA-N
15448
undecan-2-ol
C11H24O
MFCD00021958
2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
CHEBI:77930
CCCCCCCCCC(C)O

Specifications

1653-30-1
0.828
107°C (225°F)
C11H24O
MFCD00021958
2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
XMUJIPOFTAHSOK-UHFFFAOYNA-N
undecan-2-ol
15448
172.31
2-Undecanol
1°C to 3°C
228°C to 229°C
Mild
1.437
1719795
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.
CCCCCCCCCC(C)O
172.31
CHEBI:77930
≥98%
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 216-722-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only