missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanol, 98+%

Catalog No. AAA1444109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1444109 10 g
AAA1444118 50 g
AAA1444130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1444109 Supplier Thermo Scientific Chemicals Supplier No. A1444109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1653-30-1 | C11H24O | 172.31 g/mol

It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1653-30-1
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00021958
InChI Key XMUJIPOFTAHSOK-UHFFFAOYNA-N
Synonym 2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
PubChem CID 15448
ChEBI CHEBI:77930
IUPAC Name undecan-2-ol
SMILES CCCCCCCCCC(C)O

Specifications

Melting Point 1°C to 3°C
Density 0.828
Boiling Point 228°C to 229°C
Flash Point 107°C (225°F)
Odor Mild
Refractive Index 1.437
Quantity 10 g
Beilstein 1719795
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.
Formula Weight 172.31
Percent Purity ≥98%
Chemical Name or Material 2-Undecanol
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 216-722-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only