missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanol, 98+%

CAS: 1653-30-1 | C11H24O | 172.31 g/mol

Supplier:  Thermo Scientific Chemicals A1444118

Quantity 50 g

 View more versions of this product

Catalog No. AAA1444118


Only null left

Description

Description

It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

1653-30-1
172.31
XMUJIPOFTAHSOK-UHFFFAOYNA-N
15448
undecan-2-ol
C11H24O
MFCD00021958
2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
CHEBI:77930
CCCCCCCCCC(C)O

Specifications

2-Undecanol
1°C to 3°C
228°C to 229°C
Mild
1.437
MFCD00021958
2-undecanol, methyl nonyl carbinol, 2-hydroxyundecane, 2-hendecanol, sec-undecyl alcohol, undecylic alcohol, sec, methylnonylcarbinol, fema no. 3246, sec-undecylic alcohol, +-2-undecanol
XMUJIPOFTAHSOK-UHFFFAOYNA-N
undecan-2-ol
15448
172.31
1653-30-1
0.828
107°C (225°F)
C11H24O
50 g
1719795
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.
CCCCCCCCCC(C)O
172.31
CHEBI:77930
≥98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 216-722-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions