missing translation for 'onlineSavingsMsg'
Learn More

2-(Triphenylphosphoranylidene)propionaldehyde, 98%

Catalog No. AC399110100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399110100 10 g Glass bottle
AC399110010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399110100 Supplier Thermo Scientific Chemicals Supplier No. 399110100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24720-64-7 | C21H19OP | 318.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 24720-64-7
Molecular Formula C21H19OP
Molecular Weight (g/mol) 318.36
MDL Number MFCD00075596
InChI Key VHUQEFAWBCDBSC-UHFFFAOYSA-N
Synonym 2-triphenylphosphoranylidene propionaldehyde, 2-triphenylphosphoranylidene propanal, propanal, 2-triphenylphosphoranylidene, 1-formylethylidenetriphenylphosphorane, 2-triphenyl-??-phosphanylidene propanal, acmc-20aol9, 2-triphenylylidene propanal, 1-formylethylidene triphenyl phosphorane, alpha-formylethylidenetriphenylphosphorane, propanal,2-triphenylphosphoranylidene
PubChem CID 4280987
IUPAC Name 2-(triphenyl-$l^{5}-phosphanylidene)propanal
SMILES CC(C=O)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 213.0°C to 217.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5)3P=C(CH3)COH
Quantity 10 g
Formula Weight 318.35
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 2-(Triphenylphosphoranylidene)propionaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only