accessibility menu, dialog, popup

UserName

CH3PO

CAS RN 24720-64-7

IUPAC Name: 2-(triphenyl-λ⁵-phosphanylidene)propanal
Synonyms: 2-(triphenyl-λ⁵-phosphanylidene)propanal
Molecular Weight (g/mol): 318.36
Molecular Formula: C21H19OP
InChi Key: VHUQEFAWBCDBSC-UHFFFAOYSA-N
SMILES: CC(C=O)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results