missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. p-7136786
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14585G 5 g
T145825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14585G Supplier TCI America Supplier No. T14585G
Only null left

Chemical Identifiers

CAS 82495-75-8
Molecular Formula C24H30ClOPSi
Molecular Weight (g/mol) 429.012
MDL Number MFCD00043159
InChI Key NEUMNYXEDIPGJD-UHFFFAOYSA-M
Synonym 2-trimethylsilyl ethoxymethyltriphenylphosphonium chloride, sem-triphenylphosphonium chloride, 2-trimethylsilyl ethoxymethyl triphenylphosphonium chloride, phosphonium, triphenyl 2-trimethylsilyl ethoxy methyl-, chloride, acmc-209pp9, 2-trimethylsilyl ethoxymethyltriphenylphosphoniumchloride, 2-trimethylsilyl ethoxymethyl-triphenylphosphonium chloride, 2-trimethylsilyl-ethoxymethyl triphenylphosphonium chloride, triphenyl-2-trimethylsilanyl-ethoxymethyl phosphonium chloride
PubChem CID 11396265
IUPAC Name triphenyl(2-trimethylsilylethoxymethyl)phosphanium;chloride
SMILES C[Si](C)(C)CCOC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]

Specifications

Melting Point 165°C
Color White
Quantity 5 g
Formula Weight 429.01
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium Chloride