missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)thioxanthen-9-one 98.0+%, TCI America™
Supplier: TCI America T238425G
Specifications
| 2-(Trifluoromethyl)thioxanthen-9-one | |
| 149°C | |
| C14H7F3OS | |
| 25 g | |
| NEWRXGDGZGIHIS-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)-9H-thioxanthen-9-one | |
| 74324 | |
| ≥98.0% (GC) |
| 1693-28-3 | |
| Yellow | |
| MFCD00079785 | |
| 2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, 2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, pubchem10678, acmc-209dzj | |
| FC(F)(F)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1 | |
| 280.26 | |
| 280.26 | |
| Crystalline Powder |
Chemical Identifiers
| 1693-28-3 | |
| 280.26 | |
| NEWRXGDGZGIHIS-UHFFFAOYSA-N | |
| 74324 | |
| FC(F)(F)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1 |
| C14H7F3OS | |
| MFCD00079785 | |
| 2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, 2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, pubchem10678, acmc-209dzj | |
| 2-(trifluoromethyl)-9H-thioxanthen-9-one |
Safety and Handling
TSCA : No