missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)thioxanthen-9-one 98.0+%, TCI America™

Catalog No. p-7137190
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23845G 5 g
T238425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23845G Supplier TCI America Supplier No. T23845G
Only null left

Chemical Identifiers

CAS 1693-28-3
Molecular Formula C14H7F3OS
Molecular Weight (g/mol) 280.26
MDL Number MFCD00079785
InChI Key NEWRXGDGZGIHIS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl thioxanthen-9-one, 2-trifluoromethyl thioxanthone, 2-trifluoromethyl-9h-thioxanthen-9-one, 2-trifluoromethylthioxanthone, 9h-thioxanthen-9-one, 2-trifluoromethyl, 2-trifludromethylthioxanthone, 2-trifluormethyl-9-thiaxanthen-9-one, 2-trifluoromethyl dibenzo b,e thiin-10-one, pubchem10678, acmc-209dzj
PubChem CID 74324
IUPAC Name 2-(trifluoromethyl)-9H-thioxanthen-9-one
SMILES FC(F)(F)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 149°C
Color Yellow
Quantity 5 g
Formula Weight 280.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)thioxanthen-9-one