accessibility menu, dialog, popup

UserName

CAS RN 1693-28-3

IUPAC Name: 2-(trifluoromethyl)-9H-thioxanthen-9-one
Synonyms: 2-(trifluoromethyl)thioxanthen-9-one
Molecular Weight (g/mol): 280.26
Molecular Formula: C14H7F3OS
InChi Key: NEWRXGDGZGIHIS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C2SC3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results