Learn More
2-(Trifluoromethyl)pyrrolidine, 97%, Thermo Scientific™
$1,189.82
Chemical Identifiers
| CAS | 109074-67-1 |
|---|---|
| Molecular Formula | C5H8F3N |
| Molecular Weight (g/mol) | 139.12 |
| InChI Key | VINAMCOZNJHNIH-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole |
| PubChem CID | 2782839 |
| IUPAC Name | 2-(trifluoromethyl)pyrrolidine |
| SMILES | C1CC(NC1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC454810050
|
N/A | 5g |
Each for $1,189.82
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 109074-67-1 | |
| 139.12 | |
| 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole | |
| 2-(trifluoromethyl)pyrrolidine |
| C5H8F3N | |
| VINAMCOZNJHNIH-UHFFFAOYSA-N | |
| 2782839 | |
| C1CC(NC1)C(F)(F)F |
Specifications
| 109074-67-1 | |
| 110.0°C to 112.0°C | |
| Authentic | |
| C5H8F3N | |
| 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole | |
| C1CC(NC1)C(F)(F)F | |
| 139.12 | |
| 139.12 | |
| 2-(Trifluoromethyl)pyrrolidine |
| 1.2100g/mL | |
| 26°C | |
| 96% min. (GC) | |
| 1.21 | |
| VINAMCOZNJHNIH-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)pyrrolidine | |
| 2782839 | |
| 97% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to
GHS Signal Word: Warning
RUO – Research Use Only