missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyrrolidine, 97%, Thermo Scientific Chemicals

Catalog No. AC454810050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454810050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454810050 Supplier Thermo Scientific Chemicals Supplier No. 454810050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109074-67-1 | C5H8F3N | 139.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109074-67-1
Molecular Formula C5H8F3N
Molecular Weight (g/mol) 139.12
InChI Key VINAMCOZNJHNIH-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole
PubChem CID 2782839
IUPAC Name 2-(trifluoromethyl)pyrrolidine
SMILES C1CC(NC1)C(F)(F)F

Specifications

Density 1.2100g/mL
Boiling Point 110.0°C to 112.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.21
Formula Weight 139.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyrrolidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to

RUO – Research Use Only