accessibility menu, dialog, popup

UserName

NHFFF

CAS RN 109074-67-1

IUPAC Name: 2-(trifluoromethyl)pyrrolidine
Synonyms: 2-(trifluoromethyl)pyrrolidine
Molecular Weight (g/mol): 139.12
Molecular Formula: C5H8F3N
InChi Key: VINAMCOZNJHNIH-UHFFFAOYNA-N
SMILES: FC(F)(F)C1CCCN1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)