Learn More
2-(Trifluoromethyl)pyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6331503
Description
2-(Trifluoromethyl)pyridine-4-carboxylic acid, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Trifluoromethyl)pyridine-4-carboxylic acid, is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
| 2-(Trifluoromethyl)pyridine-4-carboxylic acid | |
| 217°C to 223°C | |
| C7H4F3NO2 | |
| 1 g | |
| Slightly soluble in water. | |
| OC(=O)C1=CC(=NC=C1)C(F)(F)F | |
| 191.11 | |
| 191.11 | |
| Solid |
| 131747-41-6 | |
| White | |
| MFCD07774132 | |
| 2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl | |
| BZFGKBQHQJVAHS-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)pyridine-4-carboxylic acid | |
| 14761447 | |
| 97% |
Chemical Identifiers
| 131747-41-6 | |
| 191.11 | |
| BZFGKBQHQJVAHS-UHFFFAOYSA-N | |
| 14761447 | |
| OC(=O)C1=CC(=NC=C1)C(F)(F)F |
| C7H4F3NO2 | |
| MFCD07774132 | |
| 2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl | |
| 2-(trifluoromethyl)pyridine-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only