accessibility menu, dialog, popup

UserName

NOHOFFF

CAS RN 131747-41-6

IUPAC Name: 2-(trifluoromethyl)pyridine-4-carboxylic acid
Synonyms: 2-(trifluoromethyl)pyridine-4-carboxylic acid
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: BZFGKBQHQJVAHS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)