missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7054883
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63315MD 250 mg
AAH6331503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63315MD Supplier Thermo Scientific Chemicals Supplier No. H63315MD

2-(Trifluoromethyl)pyridine-4-carboxylic acid, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyridine-4-carboxylic acid, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 131747-41-6
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD07774132
InChI Key BZFGKBQHQJVAHS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl
PubChem CID 14761447
IUPAC Name 2-(trifluoromethyl)pyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=NC=C1)C(F)(F)F

Specifications

Melting Point 217°C to 223°C
Color White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 191.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-(Trifluoromethyl)pyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only