missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T18001G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18001G Supplier TCI America Supplier No. T18001G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1423-27-4
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.928
MDL Number MFCD00236059
InChI Key JNSBEPKGFVENFS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylboronic acid, 2-trifluoromethylphenylboronic acid, 2-trifluoromethyl phenyl boronic acid, 2-trifluoromethyl benzeneboronic acid, 2-trifluoromethyl phenyl boranediol, o-trifluoromethylphenylboronic acid, 2-boronobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolueneboronic acid
PubChem CID 2734387
IUPAC Name [2-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1C(F)(F)F)(O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 189.93
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)