accessibility menu, dialog, popup

UserName

CAS RN 1423-27-4

IUPAC Name: [2-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 189.93
Molecular Formula: C7H6BF3O2
InChi Key: JNSBEPKGFVENFS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results