missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)phenylacetonitrile 98.0+%, TCI America™
Supplier: TCI America T236625G
Specifications
| 2-(Trifluoromethyl)phenylacetonitrile | |
| White-Yellow | |
| C9H6F3N | |
| 25 g | |
| 2-trifluoromethyl phenylacetonitrile, 2-2-trifluoromethyl phenyl acetonitrile, 2-trifluoromethyl benzyl cyanide, o-trifluoromethylbenzyl cyanide, 2-trifluoromethylbenzyl cyanide, benzeneacetonitrile, 2-trifluoromethyl, o-trifluoromethylphenylacetonitrile, 2-trifluoromethyl phenyl acetonitrile | |
| FC(F)(F)C1=CC=CC=C1CC#N | |
| 185.15 | |
| 185.15 | |
| Crystalline Powder |
| 3038-47-9 | |
| 92°C | |
| MFCD00001903 | |
| 3439 | |
| QXDCZSJGEUSERL-UHFFFAOYSA-N | |
| 2-[2-(trifluoromethyl)phenyl]acetonitrile | |
| 76435 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3038-47-9 | |
| 185.15 | |
| QXDCZSJGEUSERL-UHFFFAOYSA-N | |
| 76435 | |
| FC(F)(F)C1=CC=CC=C1CC#N |
| C9H6F3N | |
| MFCD00001903 | |
| 2-trifluoromethyl phenylacetonitrile, 2-2-trifluoromethyl phenyl acetonitrile, 2-trifluoromethyl benzyl cyanide, o-trifluoromethylbenzyl cyanide, 2-trifluoromethylbenzyl cyanide, benzeneacetonitrile, 2-trifluoromethyl, o-trifluoromethylphenylacetonitrile, 2-trifluoromethyl phenyl acetonitrile | |
| 2-[2-(trifluoromethyl)phenyl]acetonitrile |
Safety and Handling
TSCA : No