missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)phenylacetonitrile 98.0+%, TCI Americaâ„¢
$64.25 - $194.65
Chemical Identifiers
CAS | 3038-47-9 |
---|---|
Molecular Formula | C9H6F3N |
Molecular Weight (g/mol) | 185.15 |
MDL Number | MFCD00001903 |
InChI Key | QXDCZSJGEUSERL-UHFFFAOYSA-N |
Synonym | 2-trifluoromethyl phenylacetonitrile, 2-2-trifluoromethyl phenyl acetonitrile, 2-trifluoromethyl benzyl cyanide, o-trifluoromethylbenzyl cyanide, 2-trifluoromethylbenzyl cyanide, benzeneacetonitrile, 2-trifluoromethyl, o-trifluoromethylphenylacetonitrile, 2-trifluoromethyl phenyl acetonitrile |
PubChem CID | 76435 |
IUPAC Name | 2-[2-(trifluoromethyl)phenyl]acetonitrile |
SMILES | FC(F)(F)C1=CC=CC=C1CC#N |
Chemical Identifiers
3038-47-9 | |
185.15 | |
QXDCZSJGEUSERL-UHFFFAOYSA-N | |
76435 | |
FC(F)(F)C1=CC=CC=C1CC#N |
C9H6F3N | |
MFCD00001903 | |
2-trifluoromethyl phenylacetonitrile, 2-2-trifluoromethyl phenyl acetonitrile, 2-trifluoromethyl benzyl cyanide, o-trifluoromethylbenzyl cyanide, 2-trifluoromethylbenzyl cyanide, benzeneacetonitrile, 2-trifluoromethyl, o-trifluoromethylphenylacetonitrile, 2-trifluoromethyl phenyl acetonitrile | |
2-[2-(trifluoromethyl)phenyl]acetonitrile |
Specifications
3038-47-9 | |
92°C | |
MFCD00001903 | |
3439 | |
QXDCZSJGEUSERL-UHFFFAOYSA-N | |
2-[2-(trifluoromethyl)phenyl]acetonitrile | |
76435 | |
≥98.0% (GC) | |
2-(Trifluoromethyl)phenylacetonitrile |
White-Yellow | |
C9H6F3N | |
5 g | |
2-trifluoromethyl phenylacetonitrile, 2-2-trifluoromethyl phenyl acetonitrile, 2-trifluoromethyl benzyl cyanide, o-trifluoromethylbenzyl cyanide, 2-trifluoromethylbenzyl cyanide, benzeneacetonitrile, 2-trifluoromethyl, o-trifluoromethylphenylacetonitrile, 2-trifluoromethyl phenyl acetonitrile | |
FC(F)(F)C1=CC=CC=C1CC#N | |
185.15 | |
185.15 | |
Crystalline Powder |
Safety and Handling
TSCA : No