missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenyl Isocyanate 98.0+%, TCI America™

Catalog No. I041210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I041210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I041210G Supplier TCI America Supplier No. I041210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2285-12-3
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.12
MDL Number MFCD00002008
InChI Key GZWGTVZRRFPVAS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
PubChem CID 16794
IUPAC Name 1-isocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=O

Specifications

Color Yellow
Boiling Point 79°C
Quantity 10 g
UN Number 2285
Formula Weight 187.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)phenyl Isocyanate
RTECSNumber NR0201000
TSCA No
Recommended Storage Refrigerator