accessibility menu, dialog, popup

UserName

FFFNCO

CAS RN 2285-12-3

IUPAC Name: 1-isocyanato-2-(trifluoromethyl)benzene
Synonyms: 1-isocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H4F3NO
InChi Key: GZWGTVZRRFPVAS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1N=C=O
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)