missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenyl isocyanate, 97%

CAS: 2285-12-3 | C8H4F3NO | 187.12 g/mol

Supplier:  Thermo Scientific Chemicals L1115606

 View more versions of this product

Catalog No. AAL1115606


Only null left
Explore available promotions

Description

Description

2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
Specifications

Specifications

2-(Trifluoromethyl)phenyl isocyanate
1.238
58°C (136°F)
C8H4F3NO
MFCD00002008
UN2285
Moisture sensitive
Hydrolyzes in water.
C1=CC=C(C(=C1)C(F)(F)F)N=C=O
187.12
187.12
2285-12-3
74°C to 75°C (40 mmHg)
Pungent
1.4755
5 g
977056
2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
GZWGTVZRRFPVAS-UHFFFAOYSA-N
1-isocyanato-2-(trifluoromethyl)benzene
16794
97%

Chemical Identifiers

2285-12-3
187.12
GZWGTVZRRFPVAS-UHFFFAOYSA-N
16794
C1=CC=C(C(=C1)C(F)(F)F)N=C=O
C8H4F3NO
MFCD00002008
2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
1-isocyanato-2-(trifluoromethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H226-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c

H226-H312-H315-H319-H330-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 218-925-5

RTECSNumber : NR0201000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only