missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenyl isocyanate, 97%

Catalog No. AAL1115603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1115603 1 g
AAL1115606 5 g
AAL1115614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1115603 Supplier Thermo Scientific Chemicals Supplier No. L1115603

CAS: 2285-12-3 | C8H4F3NO | 187.12 g/mol

2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture. Store under dry inert gas.

Chemical Identifiers

CAS 2285-12-3
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.12
MDL Number MFCD00002008
InChI Key GZWGTVZRRFPVAS-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
PubChem CID 16794
IUPAC Name 1-isocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=O

Specifications

Density 1.238
Boiling Point 74°C to 75°C (40 mmHg)
Flash Point 58°C (136°F)
Odor Pungent
Refractive Index 1.4755
Quantity 1 g
UN Number UN2285
Beilstein 977056
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 187.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)phenyl isocyanate
Show More Show Less
Hazard Category H226-H312-H315-H319-H330-H334-H335
Hazard Statement GHS H Statement
H330-H334-H226-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 218-925-5
RTECSNumber NR0201000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only