missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)isonicotinic Acid 98.0+%, TCI America™
Supplier: TCI America T32691G
Specifications
| 2-(Trifluoromethyl)isonicotinic Acid | |
| 221°C | |
| C7H4F3NO2 | |
| 1 g | |
| BZFGKBQHQJVAHS-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)pyridine-4-carboxylic acid | |
| 14761447 | |
| ≥98.0% (GC,T) |
| 131747-41-6 | |
| White-Yellow | |
| MFCD07774132 | |
| 2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl | |
| OC(=O)C1=CC(=NC=C1)C(F)(F)F | |
| 191.11 | |
| 191.11 | |
| Crystalline Powder |
Chemical Identifiers
| 131747-41-6 | |
| 191.11 | |
| BZFGKBQHQJVAHS-UHFFFAOYSA-N | |
| 14761447 | |
| OC(=O)C1=CC(=NC=C1)C(F)(F)F |
| C7H4F3NO2 | |
| MFCD07774132 | |
| 2-trifluoromethyl isonicotinic acid, 2-trifluoromethyl pyridine-4-carboxylic acid, 2-trifluoromethyl-isonicotinic acid, 2-trifluoromethyl isonicotinicacid, 4-pyridinecarboxylic acid, 2-trifluoromethyl, 2-trifluoromethyl-4-pyridinecarboxylic acid, pubchem18487, acmc-1c8b9, ksc174g3f, 4-pyridinecarboxylicacid, 2-trifluoromethyl | |
| 2-(trifluoromethyl)pyridine-4-carboxylic acid |
Safety and Handling
TSCA : No