missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzyl Chloride 98.0+%, TCI America™

Catalog No. p-7137379
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27315G 5 g
T273125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27315G Supplier TCI America Supplier No. T27315G

Chemical Identifiers

CAS 21742-00-7
Molecular Formula C8H6ClF3
Molecular Weight (g/mol) 194.581
MDL Number MFCD00009920
InChI Key BBXDMCQDLOCXRA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzyl chloride, 1-chloromethyl-2-trifluoromethyl benzene, 2-trifluoromethyl benzylchloride, 2-trifluoromethylbenzyl chloride, o-trifluoromethylbenzyl chloride, benzene, 1-chloromethyl-2-trifluoromethyl, alpha'-chloro-alpha,alpha,alpha-trifluoro-o-xylene, g1r bxfff, pubchem4692, acmc-209fnp
PubChem CID 89028
IUPAC Name 1-(chloromethyl)-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)C(F)(F)F

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
UN Number 3265
Formula Weight 194.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzyl Chloride