missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)benzyl Bromide 97.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Boiling Point | 72°C |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 239.04 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 2-(Trifluoromethyl)benzyl Bromide |
Safety and Handling
| TSCA | No |