accessibility menu, dialog, popup

UserName

FFFBr

CAS RN 395-44-8

IUPAC Name: 1-(bromomethyl)-2-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.04
Molecular Formula: C8H6BrF3
InChi Key: TXVVVEUSVBLDED-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results